Thermo Scientific&trade; Oxoid&trade; <i>Brilliance</i>&trade; Listeria Agar (ISO) Supplements
Thermo Scientific™
Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Supplements
Enable the performance of Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) with these selective and differential supplements.
Catalog NumberYieldForm
SR0257E1 vial / 500 mL agarPellets
SR0258E1 vial / 500 mL agarLiquid
SR0257B1 vial/ 200 mL BO1370Z mediumPellets
SR0258B1 vial/ 200 mL BO1370Z mediumLiquid
Catalog number SR0257E
Price (JPY)
-
Yield:
1 vial / 500 mL agar
Form:
Pellets
Enable the performance of Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Base with the Thermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Supplements.
When used to supplement the Brilliance™ Listeria Agar (ISO) Supplements, the Selective Supplement enables the inhibition of competing microflora whilst the Differential Supplement aids the differentiation of Listeria monocytogenes from other Listeria species by facilitating the development of an opaque white halo around the colonies.
Enable the performance of the Thermo Scientific™ Oxoid™ Brilliance™ Agar (ISO) Base with the Brilliance™  Listeria Agar (ISO) Supplements.
  • Available in convenient sizes suitable for supplementing 5 L of Brilliance Listeria Agar (ISO) (E pack size) or compatible with our 500 mL bottled media options for preparing Pour plates (B pack size).
  • Each box of supplement contains 10 vials providing a convenient way to make as much or as little Brilliance Listeria Agar (ISO) as you need.
  • Brilliance Listeria Agar (ISO) is intended to be used as part of the Thermo Scientific™ Oxoid™ Listeria Precis™ Methods or  as part of the ISO 11290-1:2017 and ISO 11290-2:2017 reference methods for the detection and enumeration of Listeria species and L. monocytogenes.
  • The Listeria Precis Methods are validated for a broad range of samples according to ISO 16140-2 following NF VALIDATION rules when compared to the ISO 11290-1:2017 and ISO 11290-2:2017: Microbiology of the food chain – Horizontal methods for the detection and enumeration of Listeria monocytogenes and of Listeria spp.
Specifications
DescriptionThermo Scientific™ Oxoid™ Brilliance™ Listeria Agar (ISO) Selective Supplement
FormatBox of 10 vials
FormulationNalidixic Acid, Polymyxin B, Amphotericin, Ceftazidime
Yield1 vial / 500 mL agar
FormPellets
Product TypeMedia Supplement
Unit SizeEach