You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Chemicals
▸
Solvents
▸
Ethanol
▸
80% Ethanol, Molecular Biology Grade
Element Search
Structure Search
Thermo Scientific™
80% Ethanol, Molecular Biology Grade
Ready made ethanol/water blend prequalified for molecular biology research applications. | CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
Change view
Quantity:
4 L
Packaging:
Poly Bottle
Unit Size:
Case of 4
Each
Try the chem dex
TM
web app
Catalog number
T08204K7CS
also known as T08204K7
Price (TWD)
-
Request A Quote
Quantity:
4 L
Packaging:
Poly Bottle
Unit Size:
Case of 4
Request bulk or custom format
Chemical Identifiers
CAS
64-17-5
IUPAC Name
ethanol
Molecular Formula
C2H6O
InChI Key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES
CCO
View more
Specifications
Specification Sheet
Assay of water
Inclusive Between 18 - 22%
Protease activity
Pass Test
Substances Darkened by H2SO4
Pass Test
DNase
Pass Test
RNase
Pass Test
View more
80% ethanol and water blend
DNase-, RNase-, and protease-free
Ready made ethanol/water blend
Pre-qualified for molecular biology research applications
Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins
Tested for use with Applied Biosystems™ MagMAX™ kits
For Manufacturing and Laboratory Use. Not for beverage use.