The red-fluorescent tetramethylrhodamine (TAMRA) azide can be reacted with terminal alkynes via a copper-catalyzed click reaction. Click chemistry describes aRead more
Have Questions?
Catalog Number
Quantity
T10182
0.5 mg
Catalog number T10182
Price (USD)
700.00
0.5 mg
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
700.00
0.5 mg
The red-fluorescent tetramethylrhodamine (TAMRA) azide can be reacted with terminal alkynes via a copper-catalyzed click reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAlkyne
Detection MethodFluorescence
FormatSolid
Label or DyeTAMRA™ Isomers, TMR (Tetramethylrhodamine)
Molecular Weight (g/mol)554.65 Da
Product TypeTetramethylrhodamine Azide
Quantity0.5 mg
Reactive GroupAzide
Reactive MoietyAmine, Azide
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
ColorOrange
Label TypeClassic Dyes
Product LineMolecular Probes
Unit Size0.5 mg
Contents & Storage
Store at ≤-20°C, desiccated and protected from light.
Citations & References (3)
Citations & References
Abstract
Exploring RNA transcription and turnover in vivo by using click chemistry.
Authors:Jao CY, Salic A,
Journal:Proc Natl Acad Sci U S A
PubMed ID:18840688
We describe a chemical method to detect RNA synthesis in cells, based on the biosynthetic incorporation of the uridine analog 5-ethynyluridine (EU) into newly transcribed RNA, on average once every 35 uridine residues in total RNA. EU-labeled cellular RNA is detected quickly and with high sensitivity by using a copper ... More
Approach to profile proteins that recognize post-translationally modified histone
Authors:Li X, Kapoor TM,
Journal:J Am Chem Soc
PubMed ID:20141135
Post-translational modifications (PTMs) of histones, proteins onto which DNA is packaged, are involved in many biological processes, including transcription, recombination, and chromosome segregation. As these PTMs can be dynamic, combinatorial, and mediators of weak interactions, the comprehensive profiling of all proteins that recognize histone PTMs is a daunting task. Here ... More
Imaging the lipidome: omega-alkynyl fatty acids for detection and cellular visualization of lipid-modified proteins.
Authors:Hannoush RN, Arenas-Ramirez N,
Journal:ACS Chem Biol
PubMed ID:19505150
Fatty acylation or lipid modification of proteins controls their cellular activation and diverse roles in physiology. It mediates protein-protein and protein-membrane interactions and plays an important role in regulating cellular signaling pathways. Currently, there is need for visualizing lipid modifications of proteins in cells. Herein we report novel chemical probes ... More