Talos™ L120C TEM for Life Sciences
Talos™ L120C TEM for Life Sciences
Talos™ L120C TEM for Life Sciences
Talos™ L120C TEM for Life Sciences
Talos™ L120C TEM for Life Sciences
Thermo Scientific™
Talos™ L120C TEM for Life Sciences
Discover the next generation 120kV imaging platform. Key features of the Thermo Scientific™ Talos™ L120C transmission electron microscope (TEM) include modular design and improved optical stability, yielding unmatched ease of use, productivity, operational comfort, and uptime.
Have Questions?
Catalog number TTALOSL120C
Price (USD)/ Each
-
Request A Quote
Find additional information here: Thermo Scientific™ Talos™ L120C transmission electron microscope (TEM)
Discover the next generation 120kV imaging platform. Key features of the Thermo Scientific™ Talos™ L120C transmission electron microscope (TEM) include modular design and improved optical stability, yielding unmatched ease of use, productivity, operational comfort, and uptime. The Talos L120C TEM is an ideal solution for entry level cryo-studies, simple point and click imaging, and a highly efficient instrument for tomography and single particle analysis sample screening with standalone options for EDS and STEM.
Benefits of the Talos L120C TEM for Life Sciences
More stability.  Robust system enclosure, constant power lenses, and remote operation allow consistent use. 
Automation.  Multiple auto-functions (auto-gun, auto-alignment) improve repeatability and reproducibility of results. 
High-quality imaging.  4k × 4K Ceta CMOS camera provides a large field-of-view and live digital zooming with high sensitivity and high speed. 
Quick sample exchange.  Robust vacuum system provides a contamination-free environment and fast recovery after airlock. 
Data correlation.  MAPS software allows for automated and unattended large-area acquisition at multiple scales.
Cryo-imaging.  Observe cryo-specimen with minimal ice growth and automate single particle analysis imaging with EPU software
Automation and Ease of Use
The Talos L120C TEM features easy-to-use software, operating on Windows™ 7, which allows for switching between TEM and STEM mode quickly. The Talos L120C TEM offers a high level of automation, allowing complete digital control over multiple microscope components, including the electron gun, optical elements, vacuum system, and stage. Additionally, the user interface allows for smart presets to save multiple operational conditions across many different applications.
Providing a single software solution, the modular MAPS software platform allows for automated and unattended-large area acquisition at multiple scales. MAPS allows for image and data transfer between multiple imaging platforms, such as TEM, SEM, SDB, and Light Microscopes. The Talos L120C TEM user is able to quickly and easily correlate data from multiple imaging platforms, ready for input into the optional Amira software for advanced visualization and analysis.
Sample Protection
An improved vacuum system reduces vacuum recovery after airlock procedure to less than one minute, resulting in quick sample exchange. This new vacuum system is also more robust, providing an optimal contamination-free environment with a contamination rate of ≤ 1·104 nm3 per minute. Integrated application software allows for observation of cryo specimens with minimal ice growth, measured at less than 0.7 nm per hour.
Ready for the Future
With the potential to address multiple applications, the Talos L120C TEM is an ideal entry-level solution for imaging and tomography, and it can be configured as a basic cryo-TEM imaging platform. Fully upgradeable and able to expand as your research requirements and ambitions grow, the Talos L120C TEM will meet your needs, whether those needs are in Cryo or Room Temperature or 2D imaging or 3D imaging and multi-modality imaging experiments.
Specifications
Product Size-
Unit SizeEach