Product Image
Thermo Scientific™
InstrumentConnect Premium Subscription
Instrument Connect is a device monitoring application within our Connect cloud-based platform. While Instrument Connect basic access is free, anWeitere Informationen
Have Questions?
Katalognummer ICPAU0005D0250
Preis (EUR)
-
Instrument Connect is a device monitoring application within our Connect cloud-based platform. While Instrument Connect basic access is free, an InstrumentConnect Plus subscription provides access to expanded equipment data, application advanced features, and additional data reporting functionality.
Instrument Connect features include:
• Link your lab equipment to InstrumentConnect
• Monitor your device health
• Receive alerts and notifications for all alarms and events
• Maintain peace of mind with 24-7 access to your instrument data from anywhere in the world
The Instrument Connect app monitors a variety of laboratory equipment. Depending on the device, various additional features and benefits are available with an Instrument Connect Plus subscription. These features are documented below for the TSX ULT Freezer:
TSX ULT Freezer: Instrument Connect Basic
• Monitoring of setpoint and cabinet temperature
• Monitoring of events and alarms from the freezer
• Generic event descriptions
• 7-day notification card of active alarms/events
• One month of historical data storage and report retrieval
TSX ULT Freezer: Instrument Connect Plus
• Monitoring of setpoint and cabinet temperature
• Monitoring of additional 12+ diagnostic refrigeration system temperatures and parameters
• Monitoring of events and alarms from the freezer
• Detailed event descriptions
• Up to 6 month notification card of active alarms/events
• Full (lifetime) historical data storage and report retrieval
• Summary health report generation (on-demand and planned)
• Customizable user dashboard
• Customizable notifications for select alarms
• Issue to resolution tracking
• Bulk download and export capability across multiple units under subscription
Learn more about the Instrument Connect app and our Connect platform ›.
Specifications
Zur Verwendung mit (Geräte)Laboratory Equipment: TSX ULT
EnthältSubscription Only
TypInstrumentConnect Plus Subscription
Unit SizeEach