Product Image
Thermo Scientific™

Ultra-Sensitive ABC Peroxidase Mouse IgG Staining Kit

Thermo Scientific Pierce ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemicalRead more
Have Questions?
Catalog NumberQuantity
320528 mL
Catalog number 32052
Price (INR)
63,474.00
온라인 행사
Ends: 31-Dec-2026
105,789.00
Save 42,315.00 (40%)
Each
Quantity:
8 mL
Request bulk or custom format
Price (INR)
63,474.00
온라인 행사
Ends: 31-Dec-2026
105,789.00
Save 42,315.00 (40%)
Each
Thermo Scientific Pierce ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemical detection with biotinylated secondary antibodies.

Pierce ABC Staining Kits are highly sensitive, produce very low background staining and have rapid avidin-biotin interactions. Standard Kits contain only the two essential reagents (avidin and biotinylated HRP) can be used with nearly any species of biotinylated secondary antibody. The Ultra-Sensitive ABC Peroxidase Staining Kits are approximately five times more sensitive than the regular-senstivity kit, enabling more dilute primary antibodies to be used while producing equal staining intensity. Ultra-Sensitive Kits are offered as a Standard Kit and as Complete Kits, which contain biotinylated secondary antibodies and appropriate block serum for use with mouse or rabbit antibodies, respectively.

Related Products
ABC Peroxidase Standard Staining Kit
Ultra-Sensitive ABC Peroxidase Standard Staining Kit
Ultra-Sensitive ABC Peroxidase Rabbit IgG Staining Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Ultrasensitive ABC Peroxidase Mouse IgG Staining Kit
Detection MethodColorimetric, Biotin-Based, Chromogenic
For Use With (Application)Immunodetection, IHC (Immunohistochemistry)
Product TypeStaining Kit
Quantity8 mL
Substrate TypeHRP (Horseradish Peroxidase) Substrate
Product LinePierce
Unit SizeEach
Contents & Storage
Sufficient For: At least 10 mL of ABC staining solution
  • Reagent A (Avidin), 2 mL
  • Reagent B (Biotinylated HRP), 2 mL
  • Blocking Serum (horse), 3 mL
  • Biotinylated Horse Anti-Mouse IgG, 1 mL
  • Mixing bottles with dropper dispenser tips, 3

    Store all reagents at 4°C
  • Citations & References (1)

    Citations & References
    Abstract
    Suppressing aberrant GluN3A expression rescues synaptic and behavioral impairments in Huntington's disease models.
    Authors:Marco S, Giralt A, Petrovic MM, Pouladi MA, Martínez-Turrillas R, Martínez-Hernández J, Kaltenbach LS, Torres-Peraza J, Graham RK, Watanabe M, Luján R, Nakanishi N, Lipton SA, Lo DC, Hayden MR, Alberch J, Wesseling JF, Pérez-Otaño I
    Journal:
    PubMed ID:23852340
    Huntington's disease is caused by an expanded polyglutamine repeat in the huntingtin protein (HTT), but the pathophysiological sequence of events that trigger synaptic failure and neuronal loss are not fully understood. Alterations in N-methyl-D-aspartate (NMDA)-type glutamate receptors (NMDARs) have been implicated. Yet, it remains unclear how the HTT mutation affects ... More