Ultra-Sensitive ABC Peroxidase Standard Staining Kit
Thermo Scientific™

Ultra-Sensitive ABC Peroxidase Standard Staining Kit

Thermo Scientific Pierce ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemicalRead more
Have Questions?
Catalog NumberQuantity
320504 mL
Catalog number 32050
Price (USD)
568.65
Online exclusive
646.00
Save 77.35
Each
Quantity:
4 mL
Request bulk or custom format
Price (USD)
568.65
Online exclusive
646.00
Save 77.35
Each
Thermo Scientific Pierce ABC Staining Kits include reagents for the avidin-biotin complex (ABC) technique, a highly sensitive method for immunohistochemical detection with biotinylated secondary antibodies.

Pierce ABC Staining Kits are highly sensitive, produce very low background staining and have rapid avidin-biotin interactions. Standard Kits contain only the two essential reagents (avidin and biotinylated HRP) can be used with nearly any species of biotinylated secondary antibody. The Ultra-Sensitive ABC Peroxidase Staining Kits are approximately five times more sensitive than the regular-senstivity kit, enabling more dilute primary antibodies to be used while producing equal staining intensity. Ultra-Sensitive Kits are offered as a Standard Kit and as Complete Kits, which contain biotinylated secondary antibodies and appropriate block serum for use with mouse or rabbit antibodies, respectively.

Related Products
ABC Peroxidase Standard Staining Kit
Ultra-Sensitive ABC Peroxidase Mouse IgG Staining Kit
Ultra-Sensitive ABC Peroxidase Rabbit IgG Staining Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Ultrasensitive ABC Standard Peroxidase Staining Kit
Detection MethodColorimetric, Biotin-Based, Chromogenic
For Use With (Application)Immunodetection, IHC (Immunohistochemistry)
Product TypeStaining Kit
Quantity4 mL
Substrate TypeHRP (Horseradish Peroxidase) Substrate
Product LinePierce
Unit SizeEach
Contents & Storage
  • Reagent A (Avidin), 2 mL
  • Reagent B (Biotinylated HRP), 2 mL
  • Mixing bottle with dropper dispenser tip, 1
  • Store all reagents at 4°C