Product Image
Thermo Scientific™

NBT Substrate Powder (nitro-blue tetrazolium chloride)

Thermo Scientific Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blotRead more
Have Questions?
Catalog NumberQuantity
340351 g
Catalog number 34035
Price (INR)
22,699.00
線上優惠
Ends: 31-Dec-2026
33,627.00
Save 10,928.00 (32%)
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
22,699.00
線上優惠
Ends: 31-Dec-2026
33,627.00
Save 10,928.00 (32%)
Each
Thermo Scientific Pierce NBT and BCIP Substrates are powders and ready-made solutions of nitro-blue tetrazolium and 5-bromo-4-chloro-3'-indolyphosphate for chromogenic blot and IHC staining with alkaline phosphatase probes.

The combination of NBT (nitro-blue tetrazolium chloride) and BCIP (5-bromo-4-chloro-3'-indolyphosphate p-toluidine salt) yields an intense, insoluble black-purple precipitate when reacted with alkaline phosphatase, a popular enzyme conjugate for antibody probes. Pierce NBT and BCIP powders are available individually and in two convenient, ready-to-use solutions (with and without a levamisole suppressor of endogenous phosphatase activity).

Features:

• AP substrate—for detection of alkaline phosphatase (alk-phos) activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples
• Chromogenic—no special equipment needed for visualization; produces purple-black precipitate that is easy to photograph
• Package options—choose individual NBT and BCIP powders, or preformulated, ready-to-use solutions with or without levamisole suppressor

Highlights of Pierce NBT/BCIP 1-Step Solution (Part No. 34042):
• Best for immunoblotting applications
• Ready-to-use single component
• Sensitive with low background and minimial assay-to-assay variability

Highlights of Pierce NBT/BCIP plus Suppressor 1-Step Solution (Part No. 34070):
• Best for immunohistochemistry applications
• Ready-to-use single component
• Contains 1 mM levamisole to inhibit endogenous phosphatase activity, while yielding an intense dark purple precipitate in the presence of calf intestinal phosphatase

Related Products
1-Step™ NBT/BCIP Substrate Solution
1-Step™ NBT/BCIP plus Suppressor Substrate Solution
BCIP Substrate Powder (5-bromo-4-chloro-3'-indolyphosphate p-toluidine)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce NBT Substrate
ReactivityAlkaline Phosphatase
Detection MethodColorimetric
FormPowder
Membrane CompatibilityNitrocellulose, PVDF
Product TypeSubstrate
Quantity1 g
SubstrateAlkaline Phosphatase
Unit SizeEach
Contents & Storage
Upon receipt store product at 4°C protected from moisture. Product is shipped at ambient temperature.