Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis)
Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis)
Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis)
Thermo Scientific Chemicals

Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis)

CAS: 32287-65-3 | AlF3H2O | 101.99 g/mol
Catalog NumberQuantity
010623.06
also known as 010623-06
5 g
Catalog number 010623.06
also known as 010623-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS32287-65-3
IUPAC Nametrifluoroalumane hydrate
Molecular FormulaAlF3H2O
InChI KeyXTXFDUABTPNTFB-UHFFFAOYSA-K
SMILESO.F[Al](F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only