Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)
Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)

CAS: 1313-96-8 | Nb2O5 | 265.81 g/mol
Quantity:
50 g
250 g
1 kg
Catalog number 010822.18
also known as 010822-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS1313-96-8
IUPAC Name[(dioxoniobio)oxy]dioxoniobium
Molecular FormulaNb2O5
InChI KeyZKATWMILCYLAPD-UHFFFAOYSA-N
SMILESO=[Nb](=O)O[Nb](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Impurity contentTantalum, Ta: ≤10ppm
Total Metal Impurities≤0.0015% (≤15ppm)
Niobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products, and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators, including linear and non-linear optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only