Cerium(III) chloride hydrate, REacton™, 99.9% (REO)
Cerium(III) chloride hydrate, REacton™, 99.9% (REO)
Cerium(III) chloride hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Cerium(III) chloride hydrate, REacton™, 99.9% (REO)

CAS: 19423-76-8 | CeCl3 | 246.47 g/mol
Quantity:
50 g
250 g
Catalog number 011325.18
also known as 011325-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS19423-76-8
IUPAC Namecerium(3+) trichloride
Molecular FormulaCeCl3
InChI KeyVYLVYHXQOHJDJL-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ce+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Cerium(III) chloride hydrate is used as an important raw material the preparation of other cerium salts such as cerium(III) trifluoromethanesulfonate. It is used in the conversion of esters to allylsilanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only