Molybdenum rod, 5mm (0.2in) dia, Puratronic™, 99.97% (metals basis)
Molybdenum rod, 5mm (0.2in) dia, Puratronic™, 99.97% (metals basis)
Molybdenum rod, 5mm (0.2in) dia, Puratronic™, 99.97% (metals basis)
Thermo Scientific Chemicals

Molybdenum rod, 5mm (0.2in) dia, Puratronic™, 99.97% (metals basis)

CAS: 7439-98-7 | Mo | 95.95 g/mol
Quantity:
150 mm
10 x 150 mm
Catalog number 011475.DO
also known as 011475-DO
Price (JPY)
-
Quantity:
150 mm
Request bulk or custom format
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
FormRod, 5mm (0.2in) diameter.
Assay from Supplier's CofA≥99.97% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only