Ammonium perchlorate, Reagent Grade, Thermo Scientific Chemicals
Ammonium perchlorate, Reagent Grade, Thermo Scientific Chemicals
Ammonium perchlorate, Reagent Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium perchlorate, Reagent Grade, Thermo Scientific Chemicals
Quantity:
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011658.A1
Price (JPY)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7790-98-9
SpecificationsSpecification SheetSpecification Sheet
Insoluble material0.005% max
Impurity contentFe 0.0005% max
Impurity contentChloride + Chlorate 0.003% max
Impurity contentSO₄ 0.001% max
Trace Metals0.0005% max
View more
Ammonium perchlorate is used to make rocket propellants, explosives, pyrotechnics, as an etching and engraving agent, and in analytical chemistry.