Platinum Rhodium Palladium gauze, 80 mesh knitted from 0.076mm (0.003in) dia wire, 99.9% (metals basis)
Thermo Scientific Chemicals

Platinum Rhodium Palladium gauze, 80 mesh knitted from 0.076mm (0.003in) dia wire, 99.9% (metals basis)

Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 012216.FF
also known as 012216-FF
Price (JPY)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangePt:Rh:Pd; 90:5:5 wt%
Chemical Name or MaterialPlatinum Rhodium Palladium gauze
MDL NumberMFCD00011179
Mesh Size80 Mesh
View more

Platinum Rhodium Palladium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis) is used as a catalyst in the preparation of nitric acid from ammonia. It is also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only