Triphenylbismuth, 98+%, Thermo Scientific Chemicals
Triphenylbismuth, 98+%, Thermo Scientific Chemicals
Triphenylbismuth, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triphenylbismuth, 98+%, Thermo Scientific Chemicals

Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012545.14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS603-33-8
IUPAC Nametriphenylbismuthane
Molecular FormulaC18H15Bi
InChI KeyZHXAZZQXWJJBHA-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)[Bi](C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (unspecified)≥98.0% ( HPLC or Elemental analysis/NMR)
Melting Point≥76.0? C
Triphenylbismuth, as an volatile organometallic used in the preparation of thin films of superconducting oxides by CVD. It is also added to the fuel of rocket as cruing agent. It acts as a solidifying catalyst for butyl hydroxyl propellants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Volatile organometallic used in the preparation of thin films of superconducting oxides by CVD
RUO – Research Use Only