Bis(cyclopentadienyl)zirconium dichloride, Thermo Scientific Chemicals
Bis(cyclopentadienyl)zirconium dichloride, Thermo Scientific Chemicals
Bis(cyclopentadienyl)zirconium dichloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(cyclopentadienyl)zirconium dichloride, Thermo Scientific Chemicals

Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012548.22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1291-32-3
IUPAC Namedichlorobis(cyclopenta-2,4-dien-1-yl)zirconium
Molecular FormulaC10H10Cl2Zr
InChI KeyQRUYYSPCOGSZGQ-UHFFFAOYSA-L
SMILESCl[Zr](Cl)(C1C=CC=C1)C1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms (non-U.S. specification)
Gravimetric analysisAssay:≥94% (U.S. specification)
Gravimetric analysisZr: 30.6.-31.8 wt% (after ignition to oxide) (U.S. specification)
Melting Point242-245°C (non-U.S. specification)
Appearance (Color)White to cream
View more
Bis(cyclopentadienyl)zirconium dichloride is used for for Stereoselective Glycosidation. It is also used in the catalyzed transamidation of primary amides with amines. Zirconocene dichloride can also be used to prepare the Negishi reagent,in oxidative cyclisation reactions. Other applications include it is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only