Sodium hexahydroxyplatinate(IV), 99.9% (metals basis)
Sodium hexahydroxyplatinate(IV), 99.9% (metals basis)
Sodium hexahydroxyplatinate(IV), 99.9% (metals basis)
Thermo Scientific Chemicals

Sodium hexahydroxyplatinate(IV), 99.9% (metals basis)

CAS: 12325-31-4 | H6Na2O6Pt | 343.11 g/mol
Quantity:
1 g
5 g
Catalog number 012620.06
also known as 012620-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS12325-31-4
IUPAC Nameplatinum(4+) disodium hexahydroxide
Molecular FormulaH6Na2O6Pt
InChI KeyLDMNYTKHBHFXNG-UHFFFAOYSA-H
SMILES[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Pt+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only