Tetracyclohexyltin
Tetracyclohexyltin
Tetracyclohexyltin
Thermo Scientific Chemicals

Tetracyclohexyltin

CAS: 1449-55-4 | C24H44Sn | 451.326 g/mol
Quantity:
1 g
5 g
25 g
Catalog number 018822.03
also known as 018822-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS1449-55-4
Chemical Name or MaterialTetracyclohexyltin
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECS Number215-910-5
Formula Weight451.31
View more
It is used as the ion carrier for the construction of a highly selective salicylate sensor. This sensor shows a Nernstian response to salicylate ions over a very wide concentration (1.0 × 10-7-1.0 × 10-1 M) in a pH range of 5.5-10.5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only