Pyrophosphoryl chloride, Thermo Scientific Chemicals
Pyrophosphoryl chloride, Thermo Scientific Chemicals
Pyrophosphoryl chloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrophosphoryl chloride, Thermo Scientific Chemicals

Catalog NumberQuantity
022362.14
also known as 022362-14
25 g
Catalog number 022362.14
also known as 022362-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point101°C (10mmHg)
CAS13498-14-1
Chemical Name or MaterialPyrophosphoryl chloride
ColorColorless through Yellow
DOT InformationHazard Class: 8; Packaging Group: I
View more
Pyrophosphoryl chloride serves as a reductive chlorination reagent used in the synthesis of quetiapine. It acts as a phosphorylating agent involved in nucleoside synthesis. It plays an important role in Vilsmeier formylation and glyoxylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only