Ethylenediaminetetraacetic acid tripotassium salt
Ethylenediaminetetraacetic acid tripotassium salt
Ethylenediaminetetraacetic acid tripotassium salt
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid tripotassium salt

CAS: 17572-97-3 | C10H13K3N2O8 | 406.52 g/mol
Quantity:
25 g
100 g
Catalog number 022425.14
also known as 022425-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS17572-97-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay (unspecified)> 98.0%
Ethylenediaminetetraacetic acid tripotassium salt is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors. And it is also used to study chelators, biochemicals and reagents. EDTA tripotassium salt has been used in a study to assess the safety of EDTA and HEDTA ingredients in cosmetic formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only