Pyridine, HPLC Grade, 99.5+%
Pyridine, HPLC Grade, 99.5+%
Pyridine, HPLC Grade, 99.5+%
Pyridine, HPLC Grade, 99.5+%
Thermo Scientific™

Pyridine, HPLC Grade, 99.5+%

Maximum level of impurities: Evaporation residue 4ppm, H2O 0.05% | CAS: 110-86-1 | C5H5N | 79.102 g/mol
Quantity:
1 L
4 L
4 x 1 L
Catalog number 022905.M1
also known as 022905-M1
Price (JPY)
-
Request A Quote
Quantity:
4 x 1 L
Request bulk or custom format
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at400nm: ≤0.01 AU`
FormClear liquid
Color (APHA)≤10
Assay (GC)≥ 99.5%
Water Content (Karl Fischer Titration)≤0.05%
View more
Pyridine is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only