2-Butanone, HPLC Grade, 99.5+%
2-Butanone, HPLC Grade, 99.5+%
2-Butanone, HPLC Grade, 99.5+%
Thermo Scientific Chemicals

2-Butanone, HPLC Grade, 99.5+%

CAS: 78-93-3 | C4H8O | 72.11 g/mol
Quantity:
1 L
4 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 022924.K7
also known as 022924-K7
Price (JPY)
-
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS78-93-3
IUPAC Namebutan-2-one
Molecular FormulaC4H8O
InChI KeyZWEHNKRNPOVVGH-UHFFFAOYSA-N
SMILESCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at335 nm: ≤0.5 AU
Optical Absorbance at344 nm: ≤0.05 AU
Optical Absorbance at400 nm: ≤0.01 AU
Residue on Evaporation≤0.0025%
Optical Absorbance at329 nm: ≤1 AU
View more
2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO. It is used as a mobile phase in High Performance Liquid Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only