Triethylphosphine, 97%
Triethylphosphine, 97%
Triethylphosphine, 97%
Thermo Scientific Chemicals

Triethylphosphine, 97%

CAS: 554-70-1 | C6H15P | 118.16 g/mol
Quantity:
5 g
25 g
Catalog number 030177.06
also known as 030177-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS554-70-1
IUPAC Nametriethylphosphane
Molecular FormulaC6H15P
InChI KeyRXJKFRMDXUJTEX-UHFFFAOYSA-N
SMILESCCP(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (unspecified)>97%
FormLiquid
Triethylphosphine is used as a ligand for arylation. It is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only