Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 1.6;ID (mm), 0.8
Thermo Scientific Chemicals
Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 1.6;ID (mm), 0.8
Quantity:
200 mm
300 mm
400 mm
Catalog number 031485.QAT
also known as 031485-Q@
Price (JPY)
-
Quantity:
400 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialAl-23 Insulating Tube
Name NoteBoth ends open (Thin Wall)
Recommended StorageAmbient temperatures
TSCAU
Quantity400 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.