Dichlorophenylborane, 96%
Dichlorophenylborane, 96%
Dichlorophenylborane, 96%
Thermo Scientific Chemicals

Dichlorophenylborane, 96%

CAS: 873-51-8 | C6H5BCl2 | 158.816 g/mol
Quantity:
1 g
5 g
25 g
Catalog number 032583.03
also known as 032583-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS873-51-8
IUPAC Namedichloro(phenyl)borane
Molecular FormulaC6H5BCl2
InChI KeyNCQDQONETMHUMY-UHFFFAOYSA-N
SMILESClB(Cl)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥95.0%
FormLiquid
Dichlorophenylborane is widely used as a catalyst in Antiproliferative macrolide and cell migration inhibitor lactimidomycin and in enantioselective synthesis of polypeptide segments using vinylogous Mukaiyama aldol reactions and in enantioselective Diels-Alder cycloadditions after its reaction with allo-threonine derivatives. It is a stereo selective alkylative ring opening of cyclic anhydrides and in cross-metathesis reaction of amino derivatives with olefines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only