Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)
Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)
Thermo Scientific Chemicals

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)

CAS: 17631-68-4 | C30H33EuF21O6 | 1040.52 g/mol
Quantity:
250 mg
1 g
5 g
Catalog number 033541.03
also known as 033541-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS17631-68-4
Chemical Name or MaterialTris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)europium(III)
EINECS Number241-616-1
Formula Weight1037.49
IUPAC Nameeuropium;(Z)-1,1,1,2,2,3,3-heptafluoro-6-hydroxy-7,7-dimethyloct-5-en-4-one
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only