Dimethylgermanium dichloride, 97+%
Dimethylgermanium dichloride, 97+%
Dimethylgermanium dichloride, 97+%
Thermo Scientific Chemicals

Dimethylgermanium dichloride, 97+%

CAS: 1529-48-2 | C2H6Cl2Ge | 173.6 g/mol
Quantity:
2 g
10 g
Catalog number 034107.04
also known as 034107-04
Price (JPY)
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS1529-48-2
Molecular FormulaC2H10Cl2Ge
InChI KeyPDIYLVAYJQRPJM-UHFFFAOYSA-N
SMILESCl.Cl.C[GeH2]C
Molecular Weight (g/mol)177.63
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormLiquid
Assay from Suppliers CofA≥97% (GC)
Dimethylgermanium dichloride is a useful organogermanium compound for synthesis and proteomics research. It is also used as an organic intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only