Sodium hydroxide, 5% w/v aq. soln.
Sodium hydroxide, 5% w/v aq. soln.
Sodium hydroxide, 5% w/v aq. soln.
Thermo Scientific Chemicals

Sodium hydroxide, 5% w/v aq. soln.

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Quantity:
500 mL
4 L
Catalog number 035604.K7
also known as 035604-K7
Price (JPY)
-
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA5% w/v
FormLiquid
Appearance (Color)Colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only