Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene
Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene
Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene
Thermo Scientific Chemicals

Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene

CAS: 1184-61-8 | C12H32O4Sn | 359.094 g/mol
Quantity:
5 mL
25 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036563.AP
also known as 036563-AP
Price (JPY)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1184-61-8
IUPAC Nametetrakis(propan-2-ol) tin
Molecular FormulaC12H32O4Sn
InChI KeyVUDAJANAMZGVOP-UHFFFAOYSA-N
SMILES[Sn].CC(C)O.CC(C)O.CC(C)O.CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay from Supplier's CofA≥97.5% (metals basis, neat)
Concentration (w/v)9.5-10.5%
FormLiquid
Tin(IV) isopropoxide isopropanol is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, thin films doped tin oxide nanoparticles, thin films and are widely studied as gas sensors. Tin(IV) precursor solution was prepared from tin(IV) isopropoxide dissolved in isopropyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only