Methyl Red, 0.1% w/v solution in ethanol
Methyl Red, 0.1% w/v solution in ethanol
Methyl Red, 0.1% w/v solution in ethanol
Thermo Scientific Chemicals

Methyl Red, 0.1% w/v solution in ethanol

CAS: 74886-84-3
Catalog NumberQuantity
038698.AP
also known as 038698-AP
500 mL
Catalog number 038698.AP
also known as 038698-AP
Price (JPY)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
CAS74886-84-3
Chemical Name or MaterialMethyl Red
Concentration0.1% w/v solution in ethanol
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Formula Weight269.3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use