Lithium niobium ethoxide, 99+% (metals basis), 5% w/v in ethanol
Lithium niobium ethoxide, 99+% (metals basis), 5% w/v in ethanol
Lithium niobium ethoxide, 99+% (metals basis), 5% w/v in ethanol
Thermo Scientific Chemicals

Lithium niobium ethoxide, 99+% (metals basis), 5% w/v in ethanol

CAS: 86745-52-0 | C12H30LiNbO6 | 370.212 g/mol
Quantity:
25 mL
100 mL
Catalog number 039155.AC
also known as 039155-AC
Price (JPY)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS86745-52-0
IUPAC Nameniobium(5+) lithium(1+) hexaethanolate
Molecular FormulaC12H30LiNbO6
InChI KeyAVEKRHJBCCYCED-UHFFFAOYSA-N
SMILES[Li+].[Nb+5].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Impurity contentCa: ≤3ppm
Appearance (Color)Pale yellow
Assay from Supplier's CofA≥99.0% (metals basis)
Impurity contentCr: ≤0.5ppm
View more
Lithium niobium ethoxide is used in the preparation of lithium niobate, which is a ferroelectric material used as an optical switch. It is also used as solid electrolytes due to its high ionic diffusivity in the field of material science.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Manufacturing and Laboratory Use. Not for beverage use