Zinc phthalocyanine, 95%
Zinc phthalocyanine, 95%
Zinc phthalocyanine, 95%
Thermo Scientific Chemicals

Zinc phthalocyanine, 95%

CAS: 14320-04-8 | C32H16N8Zn | 577.916 g/mol
Quantity:
2 g
10 g
50 g
Catalog number 039553.09
also known as 039553-09
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Specifications
CAS14320-04-8
ChEBICHEBI:51218
Chemical Name or MaterialZinc phthalocyanine
ColorPurple
EINECS Number238-262-5
View more
Zinc phthalocyanine finds general applications in industry as catalysts, photoconductors and in biomedical field for photodynamic therapy, PDT. ZnPc loaded PLGA nanoparticles is a promising drug delivery system for photodynamic therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only