N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS
Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 10% TMCS

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol
Quantity:
5 g
25 g
Catalog number 040174.14
also known as 040174-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Beilstein3607620
CAS25561-30-2
Chemical Name or MaterialN,O-Bis(trimethylsilyl)trifluoroacetamide
ColorColorless to Yellow
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
View more
N,O-Bis(trimethylsilyl)trifluoroacetamide is a powerful silylation reagent useful in GC and GC-MS for the derivatization of a wide range of functional groups under mild conditions. N,O. It is also used in HPLC- electrochemical detection assay of 8-oxo-deoxyguanosine and?8-oxo-guanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only