Silicon sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis)
Silicon sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis)
Silicon sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis)
Thermo Scientific Chemicals

Silicon sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis)

CAS: 7440-21-3 | Si | 28.09 g/mol
Catalog NumberQuantity
040891.KS1 Ea.
Catalog number 040891.KS
Price (JPY)
-
Quantity:
1 Ea.
Request bulk or custom format
Chemical Identifiers
CAS7440-21-3
IUPAC Namesilicon
Molecular FormulaSi
InChI KeyXUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
FormSputtering target, 50.8mm (2.0in) diameter x 6.35mm (0.250in) thick
Assay from Supplier's CofA≥99.999% (metals basis)
Appearance (Color)Grey
Silicon sputtering target in ultra high purity silicon is used in the semiconductor industry as a result of its semiconducting properties. It is used as an alloying element in the manufacture of certain alloys (e.g. ferrosilicon, an alloy of iron and silicon which is used to introduce silicon into steel and cast iron). It is also used in the manufacture of glass and computer microchips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only