Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), recrystallized, 99.9% (metals basis)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), recrystallized, 99.9% (metals basis)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), recrystallized, 99.9% (metals basis)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), recrystallized, 99.9% (metals basis)
Thermo Scientific Chemicals

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)iron(III), recrystallized, 99.9% (metals basis)

CAS: 14876-47-2 | C33H60FeO6 | 608.682 g/mol
Quantity:
1 g
5 g
25 g
Catalog number 041862.06
also known as 041862-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS14876-47-2
IUPAC Nametris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) iron
Molecular FormulaC33H60FeO6
InChI KeyWLITYJBILWOYFF-UHFFFAOYSA-N
SMILES[Fe].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Formcrystalline powder
Total Metal Impurities
Appearance (Color)Orange to red
Elemental Analysis>/= 99.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only