Hafnium tert-butoxide, 99.9% (metals basis excluding Zr), Zr< 0.5%
Hafnium tert-butoxide, 99.9% (metals basis excluding Zr), Zr< 0.5%
Hafnium tert-butoxide, 99.9% (metals basis excluding Zr), Zr< 0.5%
Thermo Scientific Chemicals

Hafnium tert-butoxide, 99.9% (metals basis excluding Zr), Zr< 0.5%

CAS: 2172-02-3 | C16H40HfO4 | 474.98 g/mol
Quantity:
2 g
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041908.09
also known as 041908-09
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis excluding Zr), Zr < 0.5%
CAS2172-02-3
Chemical Name or MaterialHafnium tert-butoxide
ColorYellow
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only