Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS
Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS
Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS
Thermo Scientific Chemicals

Dimethyl sulfoxide-d{6}, 100% (Isotopic), contains 0.03% v/v TMS

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol
Quantity:
2 Ea.
10 Ea.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042286.SA
also known as 042286-SA
Price (JPY)
-
Quantity:
2 Ea.
Request bulk or custom format
Chemical Identifiers
CAS2206-27-1
IUPAC Name(²H₃)methanesulfinyl(²H₃)methane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Formclear liquid
Assay by NMR99.95% min Isotopic Enrichment
DMSO is a common polar solvent used in NMR, PCR, as a cryoprotectant, and as a universal solvent for many compounds. Deuterated DMSO is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only