Tri(1-naphthyl)phosphine, 97%
Tri(1-naphthyl)phosphine, 97%
Tri(1-naphthyl)phosphine, 97%
Thermo Scientific Chemicals

Tri(1-naphthyl)phosphine, 97%

CAS: 3411-48-1 | C30H21P | 412.47 g/mol
Quantity:
0.5 g
2 g
10 g
50 g
Catalog number 042304.18
also known as 042304-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS3411-48-1
IUPAC Nametris(naphthalen-1-yl)phosphane
Molecular FormulaC30H21P
InChI KeyDMEUUKUNSVFYAA-UHFFFAOYSA-N
SMILESC1=CC2=CC=CC(P(C3=CC=CC4=CC=CC=C34)C3=C4C=CC=CC4=CC=C3)=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Phosphorus-31 NMRConforms to structure
Proton NMRConforms to structure
Appearance (Color)White to pale yellow
FormPowder
Purity (DSC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only