Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals
Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals

Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042583.06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2081-12-1
IUPAC Nametetrakis(2-methylpropan-2-ol) zirconium
Molecular FormulaC16H40O4Zr
InChI KeyWRMYASGYMABHCC-UHFFFAOYSA-N
SMILES[Zr].CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
Formclear liquid
Assay (unspecified)> 97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only