Zinc wire, 0.25mm (0.01in) dia, 99.95% (metals basis)
Zinc wire, 0.25mm (0.01in) dia, 99.95% (metals basis)
Zinc wire, 0.25mm (0.01in) dia, 99.95% (metals basis)
Zinc wire, 0.25mm (0.01in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Zinc wire, 0.25mm (0.01in) dia, 99.95% (metals basis)

Form: approx. 5.60g/m | CAS: 7440-66-6 | Zn | 65.38 g/mol
Quantity:
5 m
25 m
Catalog number 042703.G5
also known as 042703-G5
Price (JPY)
-
Quantity:
5 m
Request bulk or custom format
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White silver
Assay from Supplier's CofA≥99.95% (metals basis)
FormWire, 0.25mm (0.01in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only