Nickel etchant for electrodeposited nickel films, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel etchant for electrodeposited nickel films, Thermo Scientific Chemicals

High purity etchant for electrodeposited nickel films
Catalog NumberQuantity
044580.R0
also known as 044580-R0
1 qt.
Catalog number 044580.R0
also known as 044580-R0
Price (JPY)
-
Quantity:
1 qt.
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel etchant for electrodeposited nickel films
DOT InformationHazard Class: 8; Packaging Group: II
Health Hazard 1H314-H317-H335-H350
Health Hazard 2GHS H Statement
H314-H351-H317-H319
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
Causes serious eye irritation.
Health Hazard 3P201-P202-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
View more
High purity etchant for electrodeposited nickel films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the etch rate of Nickel etchant for electrodeposited nickel films at 40 degrees C?

Nickel etchant for electrodeposited nickel films has an etch rate of 50 Å/second at 40 degrees C.

How do I reduce undercutting of an etchant?

Undercutting of the etchant can be reduced by increasing the rate of stirring or agitation.

Do I need to dilute an etchant?

No, it is a ready-to-use solution. Dilution will only be necessary if you want to reduce the etch rate.

How do I reduce the etch rate of an etchant?

The etch rate of the etchant can be reduced by adding 1 part deionized water to 2 parts etchant. This will reduce the etch rate approximately 50%.

How do I increase the etch rate of an etchant?

The etch rate of the etchant will be increased by increasing the rate of stirring or agitation.