Cyclopentane, HPLC Grade
Cyclopentane, HPLC Grade
Cyclopentane, HPLC Grade
Thermo Scientific Chemicals

Cyclopentane, HPLC Grade

For HPLC and Spectrophotometry | CAS: 287-92-3 | C5H10 | 70.135 g/mol
Quantity:
250 mL
4 L
Catalog number 044814.K7
also known as 044814-K7
Price (JPY)
-
Quantity:
4 L
Request bulk or custom format
Specifications
Beilstein1900195
CAS287-92-3
ChEBICHEBI:23492
Chemical Name or MaterialCyclopentane
ColorColorless
View more
Cyclopentane is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only