Boron nitride, 99.5% (metals basis)
Boron nitride, 99.5% (metals basis)
Boron nitride, 99.5% (metals basis)
Thermo Scientific Chemicals

Boron nitride, 99.5% (metals basis)

CAS: 10043-11-5 | BN | 24.82 g/mol
Catalog NumberQuantityPackaging
044839.KG
also known as 044839-KG
1 Pc.Bag
Catalog number 044839.KG
also known as 044839-KG
Price (JPY)
-
Quantity:
1 Pc.
Packaging:
Bag
Request bulk or custom format
Chemical Identifiers
CAS10043-11-5
IUPAC Nameboranimine
Molecular FormulaBN
InChI KeyLNLSXDSWJBUPHM-UHFFFAOYSA-N
SMILESB=N
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Boron nitride is used as polymorph, cosmetics, paints, dental cements, and pencil leads. It is also used in xerographic process and laser printers as a charge leakage barrier layer of the photo drum, fillers, processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only