Boron nitride, 99.5% (metals basis), Thermo Scientific Chemicals
Boron nitride, 99.5% (metals basis), Thermo Scientific Chemicals
Boron nitride, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron nitride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog NumberQuantity
044917.KG
also known as 044917-KG
1 Pc.
Catalog number 044917.KG
also known as 044917-KG
Price (JPY)
-
Quantity:
1 Pc.
Request bulk or custom format
Chemical Identifiers
CAS10043-11-5
IUPAC Nameboranimine
Molecular FormulaBN
InChI KeyLNLSXDSWJBUPHM-UHFFFAOYSA-N
SMILESB=N
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only