Copper gauze, 60 mesh woven from 0.19mm (0.0075in) dia wire
Copper gauze, 60 mesh woven from 0.19mm (0.0075in) dia wire
Copper gauze, 60 mesh woven from 0.19mm (0.0075in) dia wire
Thermo Scientific Chemicals

Copper gauze, 60 mesh woven from 0.19mm (0.0075in) dia wire

CAS: 7440-50-8 | Cu | 63.55 g/mol
Quantity:
150 x 150 mm
300 x 300 mm
Catalog number 045142.GT
also known as 045142-GT
Price (JPY)
-
Quantity:
150 x 150 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentNo Specifications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only