(S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(bis[3,5-dimethylphenyl]phosphine), 97%
(S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(bis[3,5-dimethylphenyl]phosphine), 97%
Thermo Scientific™

(S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(bis[3,5-dimethylphenyl]phosphine), 97%

Quantity:
250 mg
1 g
Catalog number 047218.MD
also known as 047218-MD
Price (JPY)
-
Request A Quote
Quantity:
250 mg
Request bulk or custom format
Specifications
Formula Weight814.92
Molecular FormulaC{5}{0}H{5}{6}O{6}P{2}
Synonym(S)-(4,4',5,5',6,6'-Hexamethoxy[1,1'-biphenyl]-2,2'-diyl)bis(bis[3,5-dimethylphenyl]phosphane);
Percent Purity97%
Physical FormSolid
View more