Dichloromalononitrile 97%
Dichloromalononitrile 97%
Dichloromalononitrile 97%
Thermo Scientific Chemicals

Dichloromalononitrile 97%

CAS: 13063-43-9 | C3Cl2N2 | 134.947 g/mol
Catalog number 113450050
Price (JPY)
-
Request bulk or custom format
Chemical Identifiers
CAS13063-43-9
IUPAC Namedichloropropanedinitrile
Molecular FormulaC3Cl2N2
InChI KeyNCWGPYPDPWOJKC-UHFFFAOYSA-N
SMILESClC(Cl)(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to light brown
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.4310 to 1.4360 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only