2',5'-Dimethoxyacetophenone, 99%
2',5'-Dimethoxyacetophenone, 99%
2',5'-Dimethoxyacetophenone, 99%
Thermo Scientific Chemicals

2',5'-Dimethoxyacetophenone, 99%

CAS: 1201-38-3 | C10H12O3 | 180.20 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 115340050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1201-38-3
IUPAC Name1-(2,5-dimethoxyphenyl)ethan-1-one
Molecular FormulaC10H12O3
InChI KeyFAXUIYJKGGUCBO-UHFFFAOYSA-N
SMILESCOC1=CC=C(OC)C(=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to yellow-green
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5421 to 1.5441 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only