Glycinamide hydrochloride, 98%
Glycinamide hydrochloride, 98%
Glycinamide hydrochloride, 98%
Thermo Scientific Chemicals

Glycinamide hydrochloride, 98%

CAS: 1668-10-6 | C2H6N2O·HCl | 110.54 g/mol
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 120060250
Price (JPY)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1668-10-6
IUPAC Namehydrogen 2-aminoacetamide chloride
Molecular FormulaC2H7ClN2O
InChI KeyWKNMKGVLOWGGOU-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<1 % (105°C, 4 hrs)
Appearance (Color)White to beige
Infrared spectrumConforms
TLC=<1 % Glycine
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only