Gramine, 99%
Gramine, 99%
Gramine, 99%
Thermo Scientific Chemicals

Gramine, 99%

CAS: 87-52-5 | C11H14N2 | 174.25 g/mol
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 120180250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS87-52-5
IUPAC Name[(1H-indol-3-yl)methyl]dimethylamine
Molecular FormulaC11H14N2
InChI KeyOCDGBSUVYYVKQZ-UHFFFAOYSA-N
SMILESCN(C)CC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 % (On anhydrous substance)
Loss on drying=<0.5 %
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only